missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

New Quality CHROMASOLV™ Plus Acetonitrile is suitable for all applications including:Mass spectrometry, HPLC, TLC, GPC, UV, IR, GC, Pesticide Residue Analysis, and RNA/DNA Synthesis and Purification. Product filtered through a 0.2μm filter.

£200.81 - £1611.61

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15674760
View Documents Promotion Details
Honeywell-Chromasolv
34998-1L
1 L
£200.81
1 L
Please to purchase this item. Need a web account? Register with us today!
15684760
View Documents
Honeywell-Chromasolv
34998-2.5L
2.5 L
£491.46
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15604770
View Documents
Honeywell-Chromasolv
34998-6X1L
6 x 1 L
£1054.63
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15694760
View Documents Promotion Details
Honeywell-Chromasolv
34998-4X2.5L
4 x 2.5 L
£1611.61
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
Specifications

Chemical Identifiers

75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
acetonitrile
C2H3N
MFCD00001878
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CHEBI:38472
CC#N

Specifications

-48°C
75-05-8
81°C to 82°C
C2H3N
MFCD00001878
741857
WEVYAHXRMPXWCK-UHFFFAOYSA-N
acetonitrile
6342
41.05g/mol
CHROMASOLV™ Plus
HPLC-gradient/210nm ≤1mAU
0.786g/mL at 25°C
n20/D 1.344
CH3CN
72.8mmHg (20°C)
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
CC#N
41.053
CHEBI:38472
≥99.9%
Acetonitrile, for HPLC
Videos
Safety and Handling

Safety and Handling

P210-P280-P305 + P351 + P338

H225-H302 + H312 + H332-H319

missing translation for 'einecsNumber' : 200-835-2

missing translation for 'rtecsNumber' : AL7700000

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications