missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Environmental Grade, 99.7+%, Thermo Scientific Chemicals

Product Code. 11315307
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable For Environmental, Pesticide Residue, and GC Analysis

Acetonitrile is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.05%, Halogenated residue 10ng/L (GC-ECD) 0.01%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point °C to 48°C to°C to 45°C
Color Colorless
Appearance Colorless liquid
Density 0.786
Boiling Point 80°C to 82°C
Quantity 4 L
Flash Point 5°C (41°F)
Ignition Point 525°C (977°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range >99.7%
Vapor Pressure 97 hPa (73mm Hg) at 20°C
Beilstein 741857
Solubility Information Miscible with water and almost all organic solvents.
Formula Weight 41.05
Grade Environmental
Chemical Name or Material Acetonitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.