missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072775 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
500mL
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10080000

Brand: Fisher Chemical A/0620/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 500 mL
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C
Formula Weight 41.05g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.