Learn More
Acetonitrile, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1589-5ML
Additional Details : CAS Number : 75-05-8 Weight : 0.00390kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Acetonitrile | |
75-05-8 | |
Colorless | |
ACN; Methyl cyanide | |
Analytical Standard | |
82°C (1.013 hPa) |
Glass Bottle | |
5 mL | |
CH3CN | |
41.05 | |
-46°C |