missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Solvents
- Acetonitrile
- Acetonitrile, Honeywell
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Acetonitrile, Honeywell
Product Code. 15665900 Shop All Honeywell Research Chemicals Products2.5 L
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
25 L
150 kg
This item is not returnable.
View return policy
Product Code. 15665900
Brand: Honeywell Research Chemicals 600042.5L
This item is not returnable.
View return policy
Specifications
Chemical Identifiers
CAS | 75-05-8 |
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Specifications
Density | 0.786g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.16 |
Flash Point | 2°C (35.6°F) |
Ignition Point | 973°F |
Linear Formula | CH3CN |
Vapor Pressure | 72.8 mmHg (20°C) |
UN Number | UN1648 |
Beilstein | 741857 |
Formula Weight | 41.05g/mol |
Percent Purity | ≥99.5% (GC) |
Chemical Name or Material | Acetonitrile |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
-
Product Code. 15684750
-
Product Code. 15693870
-
-
Product Code. 15695900
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 12
-
Catalog No. 10675112
£48.80 / 2.50 L -
Catalog No. 10767665
£113.00 / 2.50 L -
Catalog No. 10784941
£186.00 / 2.50 L -
Catalog No. 10284580
£72.30 / 2.50 L -
Catalog No. 10458210
£119.00 / 2.50 L -
Catalog No. 10626642
£81.40 / 2.50 L -
Catalog No. 10785901
£155.00 / 2.50 L -
Catalog No. 10045421
£33.39 / 100mL -
Catalog No. 10103483
£32.70 / 100mL -
Catalog No. 10660131
£315.00 / 2.50 L -
Catalog No. 10001334
£546.00 / 2.50 L -
Catalog No. 10407440
£266.00 / 2.50 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, Honeywell >
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur