missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications
CAS | 75-05-8 |
Quantity | 2.5 L |
Flash Point | 6 °C |
Packaging | Glass Bottle |
Physical Form | Liquid |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
Percent Purity | min. 99.9 % |
Grade | Low Water |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'