missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, HPLC for Gradient Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™ Product Code.: 10500911

Acetonitrile, HPLC for Gradient Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

Product Code. 10500911
5 L, Aluminum Can
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
This item is not returnable. View return policy

Product Code. 10500911

Brand: Fisher Chemical A/0627/21

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2H3N, CAS Number-75-05-8, acetonitrile, cyanomethane, cyanure de methyl, ethanenitrile, ethyl nitrile, mecn, methanecarbonitrile, methane, cyano, methyl cyanide, methylkyanid, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 81°C
Quantity 5 L
Packaging Aluminum Can
Physical Form Liquid
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C
Molecular Weight (g/mol) 41.05
Formula Weight 41.05g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527366 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberA/0627/PB17,A/0627/60MMS,A/0627/30RSS,A/0627/25SSY,A/0627/34X,A/0627/34VP,A/0627/15X,A/0627/99SS,A/0627/17X,A/0627/25RSS,A/0627/34SS,A/0627/21,A/0627/25SS,A/0627/15,A/0627/27SS,A/0627/PB17X,A/0627/17
Lot Number2527376 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberA/0627/PB17,A/0627/60MMS,A/0627/30RSS,A/0627/25SSY,A/0627/34X,A/0627/34VP,A/0627/15X,A/0627/99SS,A/0627/17X,A/0627/25RSS,A/0627/34SS,A/0627/21,A/0627/25SS,A/0627/15,A/0627/27SS,A/0627/PB17X,A/0627/17
Lot Number2527145 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberA/0627/PB17,A/0627/60MMS,A/0627/30RSS,A/0627/25SSY,A/0627/34X,A/0627/34VP,A/0627/15X,A/0627/99SS,A/0627/17X,A/0627/25RSS,A/0627/34SS,A/0627/21,A/0627/25SS,A/0627/15,A/0627/27SS,A/0627/PB17X,A/0627/17
Lot Number2527433 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog NumberA/0627/PB17,A/0627/60MMS,A/0627/30RSS,A/0627/25SSY,A/0627/34X,A/0627/34VP,A/0627/15X,A/0627/99SS,A/0627/17X,A/0627/25RSS,A/0627/34SS,A/0627/21,A/0627/25SS,A/0627/15,A/0627/27SS,A/0627/PB17X,A/0627/17
Lot Number2526960 Certificate TypeCertificate of Analysis Date13/02/2025 Catalog NumberA/0627/PB17,A/0627/60MMS,A/0627/30RSS,A/0627/25SSY,A/0627/34X,A/0627/34VP,A/0627/15X,A/0627/99SS,A/0627/25RSS,A/0627/34SS,A/0627/21,A/0627/25SS,A/0627/15,A/0627/27SS,A/0627/17
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, HPLC for Gradient Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.