Learn More
Acetonitrile, HPLC Gradient Grade, Thermo Scientific Chemicals
£114.57 - £243.21
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13158513
|
Thermo Scientific Acros
325730010 |
1 L | Glass Bottle |
£114.57
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10217050
|
Thermo Scientific Acros
325730025 |
2.5 L | Glass Bottle |
£243.21
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
-46.0°C | |
0.7810g/mL | |
2°C | |
99.9% | |
C2H3N | |
MFCD00001878 | |
02,13; 12,450; 13,156; 15,1 | |
Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether | |
CC#N | |
41.053 | |
CHEBI:38472 | |
99.9% | |
Acetonitrile |
75-05-8 | |
81.0°C to 82.0°C | |
1.3430 to 1.3450 | |
0.781 | |
CH3CN | |
0.36 mPa.s (20°C) | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
HPLC Gradient |
Safety and Handling
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
missing translation for 'tsca' : TSCA
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.