missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for HPLC-MS
£180.00 - £407.00
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
MDL Number | 1878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10799704
|
Fisher Chemical
A/0638/15 |
1 L |
£180.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10616653
|
Fisher Chemical
A/0638/17 |
2.5 L |
£407.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetonitrile, for HPLC-MS
Chemical Identifiers
75-05-8 | |
1878 | |
6342 | |
acetonitrile |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
CHEBI:38472 | |
CC#N |
Specifications
75-05-8 | |
Colorless | |
1878 | |
CC#N | |
41.053 | |
CHEBI:38472 | |
97mbar at 20°C | |
1.42 | |
81°C |
Amber glass bottle | |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05g/mol | |
0.36cp at 20°C | |
-46°C | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.