missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
Beskrivelse
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Tekniske data
Tekniske data
| Density | 0.786g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.16 |
| Flash Point | 2°C (35.6°F) |
| Ignition Point | 973°F |
| Linear Formula | CH3CN |
| Vapor Pressure | 72.8mmHg (20°C) |
| UN Number | 1648 |
| Beilstein | 741857 |
| Solubility Information | Completely souble in water |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion