missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Honeywell Riedel-de Haën™

Product Code. 15684670
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15684670

Brand: Honeywell-Chromasolv 348814X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C (literature)
CAS Max % 100.0000%
Density 0.786 g/mL at 25 °C(lit.)
Boiling Point 81.6°C
Quantity 4 x 2.5 L
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CH3CN
UN Number UN1648
Beilstein 741857
Formula Weight 41.05
Percent Purity >99.8%
Grade Chromatography
Chemical Name or Material Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.