missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Honeywell Riedel-de Haën™
R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)
£116.74 - £824.85
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.053 |
MDL Number | MFCD00001878 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15664670
|
Honeywell-Chromasolv
34881-1L |
1 L | Glass Bottle |
£116.74
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15674670
|
Honeywell-Chromasolv
34881-2.5L |
2.5 L | Glass Bottle |
£324.58
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694670
|
Honeywell-Chromasolv
34881-6X1L |
6 x 1 L | Glass Bottle | N/A | |||||
15684670
|
Honeywell-Chromasolv
34881-4X2.5L |
4 x 2.5 L | Glass Bottle |
£824.85
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For GC headspace applications
- Meets USP, Ph. Eur. and ICH requirements
- GC headspace application tested
- Packed under inert gas for longer shelf life
Chemical Identifiers
75-05-8 | |
41.053 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
C2H3N | |
MFCD00001878 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
Specifications
-48°C (literature) | |
100.0000% | |
81.6°C | |
C2H3N | |
MFCD00001878 | |
741857 | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
Chromatography |
75-05-8 | |
0.786 g/mL at 25 °C(lit.) | |
6°C | |
CH3CN | |
UN1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.053 | |
CHEBI:38472 | |
>99.8% | |
Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC) |
Safety and Handling
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
missing translation for 'einecsNumber' : 200-835-2
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetonitrile
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.