missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, UHPLC Grade, for Gradient Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Description
- Efficiency: lot-to-lot consistency ensures reproducible results
- Low acidity/alkalinity levels avoid interactions with samples
- Peak shape: low baseline noise at 210 nm and 254 nm improves peak shape
- Speed: filtered to 0.1 μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
- Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity

Specifications
Specifications
Melting Point | -46°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 1 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 97 mbar at 20°C |
Viscosity | 0.36 cp at 20°C |
Formula Weight | 41.05 g/mol |
Grade | UHPLC |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528445 | Certificate of Analysis | 25/02/2025 | A/0650/PB17,A/0650/PB15,A/0650/PB17Z |
2528277 | Certificate of Analysis | 25/02/2025 | A/0650/PB17,A/0650/PB15,A/0650/PB17Z |
2527366 | Certificate of Analysis | 20/02/2025 | A/0650/PB17,A/0650/PB15,A/0650/PB17Z |
2527376 | Certificate of Analysis | 18/02/2025 | A/0650/PB17,A/0650/PB15,A/0650/PB17Z |
2527145 | Certificate of Analysis | 17/02/2025 | A/0650/PB17,A/0650/PB15,A/0650/PB17Z |
Safety and Handling


Product Identifier
- Acetonitrile
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acetonitrile, UHPLC Grade, for Gradient Analysis, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction