missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, UHPLC-MS

Product Code. 15329865 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 15329865

Brand: Fisher Chemical A9561

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 75-05-8 | C2H3N | 41.053 g/mol

Acetonitrile, C2H3N, CAS Number-75-05-8, cyanure de methyl, methanecarbonitrile, cyanomethane, acetonitrile, mecn, methane, cyano, methyl cyanide, ethanenitrile, methylkyanid, ethyl nitrile, 1L

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetonitrile
Melting Point -46°C
Absorbance ≤0.005 AU at 235nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm, ≤0.01 AU at 230nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.025 AU at 215nm, ≤0.03 AU at 210nm, ≤0.04 AU at 205nm, ≤0.05 AU at 200nm, ≤0.15 AU at 195nm, ≤1.00 AU at 190nm
CAS 75-05-8
Color Colorless
Density 0.781 g/cm3
Boiling Point 81°C
Quantity 1 L
Flash Point −40°C (-40°F)
Packaging Clear Borosilicate Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9% minimum by GC
Residue after Evaporation 0.8ppm max.
Molecular Formula C2H3N
Titratable Acid 0.008meq/g max.
Titratable Base 0.0006meq/g max.
Vapor Pressure 97mbar at 20°C
Viscosity 0.36 cP at 20°C
Water 0.01% max.
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Grade UHPLC-MS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.