missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone, 98%, pure

Product Code. 10080540 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
missing translation for 'unitSize'
1 L
10 L
2.5 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10080540

missing translation for 'mfr': Thermo Scientific Acros 102410010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 98-86-2 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-86-2
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 19.6°C
Color Colorless to Yellow
Density 1.0280g/mL
Boiling Point 202.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8O
Linear Formula C6H5COCH3
Refractive Index 1.5315 to 1.5340
Quantity 1 L
Beilstein 07, IV, 619
Fieser 04,5
Merck Index 15, 71
Specific Gravity 1.028
Solubility Information Solubility in water: 5.5g/L (20°C). Other solubilities: soluble in conc. h2so4,freely soluble in alcohol,chloroform,ether and,glycerol
Molecular Weight (g/mol) 120.15
Viscosity 1.8 mPa.s (20°C)
Formula Weight 120.15
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Acetophenone
Show More Show Less
compliance-icons
Product Identifier
  • Acetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.