Learn More
Acetophenone, 99%
Description
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | 19°C to 20°C |
| Density | 1.03 |
| Boiling Point | 201°C to 203°C |
| Flash Point | 76°C (169°F) |
| Linear Formula | C6H5COCH3 |
| Refractive Index | 1.5325 |
| Quantity | 10,000 mL |
| Merck Index | 14,73 |
| Solubility Information | Miscible with water,chloroform,ethanol,ether,benzene and acetone. Slightly miscible with concentrated sulfuric acid. |
| Formula Weight | 120.15 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.