Learn More
Acetoxyacetyl chloride, 97%, Thermo Scientific Chemicals
£25.70 - £75.50
Chemische Identifikatoren
CAS | 13831-31-7 |
---|---|
Molecular Formula | C4H5ClO3 |
Molecular Weight (g/mol) | 136.53 |
MDL Number | MFCD00011535 |
InChI Key | HZDNNJABYXNPPV-UHFFFAOYSA-N |
Synonym | acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920 |
PubChem CID | 26297 |
IUPAC Name | (2-chloro-2-oxoethyl) acetate |
SMILES | CC(=O)OCC(=O)Cl |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10497402
|
Thermo Scientific Acros
370070050 |
5 g | Glass bottle |
£25.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10407412
|
Thermo Scientific Acros
370070250 |
25 g | Glass bottle |
£75.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
13831-31-7 | |
136.53 | |
HZDNNJABYXNPPV-UHFFFAOYSA-N | |
26297 | |
CC(=O)OCC(=O)Cl |
C4H5ClO3 | |
MFCD00011535 | |
acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920 | |
(2-chloro-2-oxoethyl) acetate |
Spezifikation
13831-31-7 | |
100.0 | |
1.2700g/mL | |
71°C | |
96% min. (GC) | |
CH3CO2CH2COCl | |
MFCD00011535 | |
acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920 | |
CC(=O)OCC(=O)Cl | |
136.53 | |
136.53 | |
Liquid |
96.0 | |
Colorless to Yellow | |
64°C to 68°C (25.0 mmHg) | |
Authentic | |
C4H5ClO3 | |
1.4255 to 1.4275 | |
1.27 | |
HZDNNJABYXNPPV-UHFFFAOYSA-N | |
(2-chloro-2-oxoethyl) acetate | |
26297 | |
97% | |
Acetoxyacetyl chloride, 97% |
Sicherheit und Handhabung
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-542-4
- Acetoxyacetyl chloride
Signalwort
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Gefahrenhinweis
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Sicherheitshinweis
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only