missing translation for 'onlineSavingsMsg'
Learn More

Acetyl bromide, 98%, Thermo Scientific Chemicals

£24.22 - £148.74

Chemical Identifiers

CAS 506-96-7
Molecular Formula C2H3BrO
Molecular Weight (g/mol) 122.949
MDL Number MFCD00000114
InChI Key FXXACINHVKSMDR-UHFFFAOYSA-N
Synonym ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
PubChem CID 10482
IUPAC Name acetyl bromide
SMILES CC(=O)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344619
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12485.14
25 g
£24.22
25g
Please to purchase this item. Need a web account? Register with us today!
11474104
View Documents
Thermo Scientific Alfa Aesar
A12485.22
100 g
£46.53
100g
Please to purchase this item. Need a web account? Register with us today!
11484104
View Documents
Thermo Scientific Alfa Aesar
A12485.36
500 g
£148.74
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.

Solubility
Miscible with ether, chloroform, benzene and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
Specifications

Chemical Identifiers

506-96-7
122.949
FXXACINHVKSMDR-UHFFFAOYSA-N
10482
CC(=O)Br
C2H3BrO
MFCD00000114
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
acetyl bromide

Specifications

506-96-7
1.66
>75°C (167°F)
Acrid
CH3COBr
MFCD00000114
1697546
14,82
Miscible with ether,chloroform,benzene and acetone.
CC(=O)Br
122.949
122.95
Liquid
-96°C
75°C to 77°C
≥98%
C2H3BrO
1.45
UN1716
Moisture sensitive
ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
FXXACINHVKSMDR-UHFFFAOYSA-N
acetyl bromide
10482
≥98%
Acetyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETYL BROMIDE

missing translation for 'einecsNumber' : 208-061-7

missing translation for 'rtecsNumber' : AO5955000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Moisture Sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only