Learn More
Acetyl chloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
£38.70 - £276.00
Specifications
Chemical Name or Material | Acetyl chloride |
---|---|
Color | Colorless |
Name Note | 1M solution in dichloromethane |
CAS | 75-36-5, 75-09-2 |
CAS Min % | 5.44, 93.36 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10676744
|
Thermo Scientific Acros
387741000 |
100 mL | AcroSeal™ Glass bottle |
£38.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360534
|
Thermo Scientific
387748000 |
800 mL | AcroSeal™ Glass Bottle |
£276.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetyl chloride | |
1M solution in dichloromethane | |
5.44, 93.36 | |
C2H3ClO | |
MFCD00000719 | |
1.3000g/mL | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Liquid |
Colorless | |
75-36-5, 75-09-2 | |
6.64, 94.57 | |
CH3COCl | |
0.9 to 1.1M | |
15, 80 | |
Solubility in water: reacts with water. | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
1.3 |
Safety and Handling
GHS H Statement
Suspected of causing cancer if inhaled. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Reacts violently with water. Corrosive to the respiratory tract.
GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger
- Acetyl chloride, 1M solution in dichloromethane
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH071-Corrosive to the respiratory tract.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only