missing translation for 'onlineSavingsMsg'
Learn More
Acetyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11328895

Acetyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11328895
500 mL
Click to view available options
Quantity:
25 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11328895

Brand: Thermo Scientific Alfa Aesar L14013.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

Solubility
Miscible with acetone, chloroform, glacial acetic acid, petroleum ether, ether and benzene.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662Show MoreShow Less
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

Melting Point -112°C
Density 1.104
Boiling Point 50°C to 52°C
Flash Point 4°C (39°F)
Odor Pungent
Linear Formula CH3COCl
Refractive Index 1.389
Quantity 500 mL
UN Number UN1717
Beilstein 605303
Sensitivity Moisture sensitive
Merck Index 14,85
Solubility Information Miscible with acetone,chloroform,glacial acetic acid,petroleum ether,ether and benzene.
Formula Weight 78.5
Percent Purity 98%
Chemical Name or Material Acetyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253212 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog NumberL14013.0F,L14013.AP,L14013.AC
Lot Number10251738 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL14013.0F,L14013.AP,L14013.AC
Lot Number10244014 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberL14013.0F,L14013.AP,L14013.AC
Lot Number10249157 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog NumberL14013.0F,L14013.AP,L14013.AC
Lot Number10185736 Certificate TypeCertificate of Analysis Date08/05/2023 Catalog NumberL14013.0F,L14013.AP,L14013.AC
compliance-icons compliance-icons
Product Identifier
  • Acetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetyl chloride
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    9

    Minutes

    8

    Seconds

    52

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.