Learn More
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580
£18.49 - £90.35
Identifiants chimiques
CAS | 75-36-5 |
---|---|
Molecular Formula | C2H3ClO |
Molecular Weight (g/mol) | 78.50 |
MDL Number | MFCD00000719 |
InChI Key | WETWJCDKMRHUPV-UHFFFAOYSA-N |
Synonym | ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 |
PubChem CID | 6367 |
ChEBI | CHEBI:37580 |
IUPAC Name | acetyl chloride |
SMILES | CC(Cl)=O |
Code produit | Marque | Quantity | Packaging | Prix | Quantité et disponibilité | ||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Packaging | Prix | Quantité et disponibilité | ||||
10687352
|
Thermo Scientific Acros
219470250 |
25 mL | Glass bottle |
£18.49
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10308550
|
Thermo Scientific Acros
219472500 |
250 mL | Glass bottle |
£22.51
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732384
|
Thermo Scientific Acros
219470010 |
1 L | Glass bottle |
£50.25
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772954
|
Thermo Scientific Acros
219470025 |
2.5 L | Glass bottle |
£90.35
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Spécification
75-36-5 | |
100.0 | |
1.1000g/mL | |
4°C | |
99% min. (GC) | |
CH3COCl | |
MFCD00000719 | |
01,11; 04,5; 05,4; 07,3; 11,4 | |
1.1 | |
Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
99+% | |
Acetyl chloride, p.a. |
99.0 | |
-112.0°C | |
51.0°C | |
Authentic | |
C2H3ClO | |
1.3876 to 1.3896 | |
02, 173 | |
15, 80 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Liquid |
Sécurité et traitement
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-865-6
missing translation for 'rtecsNumber' : AO6390000
missing translation for 'tsca' : TSCA
- Acetyl chloride
Mention d'avertissement
- Danger
Catégorie de danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Mention de danger
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH071-Corrosive to the respiratory tract.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only