Learn More
Acetyl chloride, 99+%, Thermo Scientific Chemicals
£25.00 - £133.00
Chemical Identifiers
CAS | 75-36-5 |
---|---|
Molecular Formula | C2H3ClO |
Molecular Weight (g/mol) | 78.50 |
MDL Number | MFCD00000719 |
InChI Key | WETWJCDKMRHUPV-UHFFFAOYSA-N |
Synonym | ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 |
PubChem CID | 6367 |
ChEBI | CHEBI:37580 |
IUPAC Name | acetyl chloride |
SMILES | CC(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11399358
|
Thermo Scientific Alfa Aesar
043262.AD |
50 mL |
£25.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389358
|
Thermo Scientific Alfa Aesar
043262.AK |
250 mL |
£43.50
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421887
|
Thermo Scientific Alfa Aesar
043262.K2 |
1 L |
£133.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful an important reactant in Friedel-Crafts reactions as well as the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-36-5 | |
78.50 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
6367 | |
acetyl chloride |
C2H3ClO | |
MFCD00000719 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
CHEBI:37580 | |
CC(Cl)=O |
Specifications
75-36-5 | |
1.104 | |
4°C (39°F) | |
C2H3ClO | |
MFCD00000719 | |
Moisture sensitive | |
Miscible with acetone,chloroform,glacial acetic acid,petroleum ether,ether and benzene. | |
CC(Cl)=O | |
78.50 | |
CHEBI:37580 | |
Liquid |
°C to 112°C | |
50°C to 52°C | |
≈99% | |
1.389 | |
605303 | |
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662 | |
WETWJCDKMRHUPV-UHFFFAOYSA-N | |
acetyl chloride | |
6367 | |
78.5 | |
Acetyl chloride |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'einecsNumber' : 200-865-6
missing translation for 'rtecsNumber' : AO6390000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only