missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl Coenzyme A Trilithium salt Trihydrate, >92%, MP Biomedicals™
£2470.00
Chemical Identifiers
CAS | 75520-41-1 |
---|---|
Molecular Formula | C23H38Li3N7O17P3S |
Molecular Weight (g/mol) | 830.39 |
InChI Key | MOGXDJCKYPGZRO-JHJDYNLLSA-N |
Synonym | acetyl coenzyme a trilithium salt |
PubChem CID | 129893900 |
IUPAC Name | S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium |
SMILES | [Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O |
Description
Chemical Identifiers
75520-41-1 | |
830.39 | |
acetyl coenzyme a trilithium salt | |
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium |
C23H38Li3N7O17P3S | |
MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
129893900 | |
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O |
Specifications
75520-41-1 | |
>92% | |
acetyl coenzyme a trilithium salt | |
[Li].[Li].[Li].CC(=O)SCCNC(=O)CCNC(=O)C(C(C)(C)COP(=O)(O)OP(=O)(O)OCC1C(C(C(O1)N2C=NC3=C2N=CN=C3N)O)OP(=O)(O)O)O | |
830.39 | |
881.1 | |
Acetyl Coenzyme A Trilithium Salt Trihydrate |
White | |
C23H38Li3N7O17P3S | |
MOGXDJCKYPGZRO-JHJDYNLLSA-N | |
S-[2-[3-[[(2R)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] ethanethioate;lithium | |
129893900 | |
Powder |
Safety and Handling
Recommended Storage : Store at -20°C.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Acetyl Coenzyme A Trilithium Salt Trihydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetylcoenzyme A