missing translation for 'onlineSavingsMsg'
Learn More
Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals Product Code.: 10586173

Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals

Product Code. 10586173
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10586173

Brand: Thermo Scientific Acros 382561000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32354-50-0
Molecular Formula C12H15O2Rh
Molecular Weight (g/mol) 294.16
MDL Number MFCD00075071
InChI Key IIQPRGKBFZGPIB-LWFKIUJUSA-M
IUPAC Name λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2

Specifications

Melting Point 140.0°C
Color Yellow
Physical Form Powder
Quantity 100 mg
Solubility Information Solubility in water: insoluble.
Formula Weight 294.16
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Acetylacetonato(norbornadiene)rhodium(I)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0216568 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number382561000,382565000
compliance-icons
Product Identifier
  • Acetylacetonato(norbornadiene)rhodium(I)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetylacetonato(norbornadiene)rhodium(I), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.