missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetone, Reagent Grade, ≥99%, Honeywell™

Reagent Grade,≥99%

£24.16 - £145.74

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15668050
View Documents
Honeywell Research Chemicals
P7754-250ML
250 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15678050
View Documents
Honeywell Research Chemicals
P7754-500ML
500 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15648050
View Documents
Honeywell Research Chemicals
P7754-1L
1 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15658050
View Documents
Honeywell Research Chemicals
P7754-2.5L
2.5 L Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

123-54-6
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(C)=O

Specifications

123-54-6
Colorless
6 (at 200.00g/L)
38°C (100.4°F)
C5H8O2
MFCD00008787
741937
153.8 - 155.2g/L at 20°C in water
CC(=O)CC(C)=O
100.12
3.5 (vs air)
100.12g/mol
Acetylacetone
-23°C
0.975g/mL at 25°C
139.5°C (1013 hPa)
Characteristic
CH3COCH2COCH3
UN2310
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
CHEBI:14750
≥99%
Videos
Safety and Handling

Safety and Handling

P210-P261-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378

missing translation for 'dotInformation' : 6.1(3)/PGIII

missing translation for 'rtecsNumber' : 204-634-0

Product Identifier
  • Acetylacetone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311 + H331-Toxic in contact with skin or if inhaled.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications