Learn More
Acetylcholine Chloride, 99%, Thermo Scientific Chemicals
£19.02 - £439.19
Chemical Identifiers
CAS | 60-31-1 |
---|---|
Molecular Formula | C7H16ClNO2 |
Molecular Weight (g/mol) | 181.66 |
MDL Number | MFCD00011698 |
InChI Key | JUGOREOARAHOCO-UHFFFAOYSA-M |
Synonym | acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride |
PubChem CID | 6060 |
ChEBI | CHEBI:2417 |
IUPAC Name | 2-acetyloxyethyl(trimethyl)azanium;chloride |
SMILES | CC(=O)OCC[N+](C)(C)C.[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10337430
|
Thermo Scientific Acros
159170050 |
5 g | Glass bottle |
£19.02
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10553901
|
Thermo Scientific Acros
159171000 |
100 g | Glass bottle |
£131.66
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465365
|
Thermo Scientific Acros
159175000 |
500 g | Glass bottle |
£439.19
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-31-1 | |
181.66 | |
JUGOREOARAHOCO-UHFFFAOYSA-M | |
6060 | |
2-acetyloxyethyl(trimethyl)azanium;chloride |
C7H16ClNO2 | |
MFCD00011698 | |
acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride | |
CHEBI:2417 | |
CC(=O)OCC[N+](C)(C)C.[Cl-] |
Specifications
60-31-1 | |
100.0 | |
White | |
Authentic | |
C7H16ClNO2 | |
MFCD00011698 | |
15, 82 | |
Solubility in water: soluble in cold water. Other solubilities: decomposes by hot water or alkalies, soluble in alcohol, chloroform (18.2g/L at 23 °C), practically insoluble in ether | |
CC(=O)OCC[N+](C)(C)C.[Cl-] | |
181.66 | |
CHEBI:2417 | |
99% | |
Acetylcholine chloride |
98.5 | |
149.0°C to 152.0°C | |
1.0% max. (105°C, 3 hrs) | |
98.5% min. (on dried basis) (Argentometry) | |
CH3CO2CH2CH2N(CH3)3Cl | |
04,III,655 | |
acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride | |
JUGOREOARAHOCO-UHFFFAOYSA-M | |
2-acetyloxyethyl(trimethyl)azanium;chloride | |
6060 | |
181.66 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-468-8
- Acetylcholine chloride
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.