missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholine chloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Endogenous neurotransmitter. Acts at nicotinic and muscarinic acetylcholine receptors.
Specifications
Specifications
CAS | 60-31-1 |
Molecular Formula | C7H16ClNO2 |
Quantity | 50 mg |
Synonym | acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride |
InChI Key | JUGOREOARAHOCO-UHFFFAOYSA-M |
SMILES | CC(=O)OCC[N+](C)(C)C.[Cl-] |
IUPAC Name | 2-acetyloxyethyl(trimethyl)azanium;chloride |
Molecular Weight (g/mol) | 181.66 |
PubChem CID | 6060 |
ChEBI | CHEBI:2417 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'