missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylic acid, 99%, Thermo Scientific Chemicals

Product Code. 10640754 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
50g
This item is not returnable. View return policy

Product Code. 10640754

Brand: Thermo Scientific Acros 158180500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetylsalicylic acid, aspirin, CAS# 50-78-2, is a non-steroidal anti-inflammatory that is used widely as an analgesic and antipyretic.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Acetylsalicylic acid is a common non-steroidal anti-inflammatory (NSAID) and
• Acetylsalicylic acid works by inhibiting cyclooxygenases, reducing prostaglandin production and the release of pyrogens


Applications
• Acetylsalicylic acid is useful for research on fever, postoperative pain, blood clots, and inflammation
• Acetylsalicylic acid may also be used in the study of cancer prevention
• Intended for research use only

Literature references

Levine, G.N.; Bates, E.R.; Bittl, J.A.; Brindis, R.G.; Fihn, S.D.; Fleisher, L.A.; Granger, C.B.; Lange, R.A.; Mack, M.J.; Mauri, L.; Mehran, R.; Mukherjee, D.; Newby, L.K.; O'Gara, P.T.; Sabatine, M.S.; Smith, P.K.; Smith, S.C. Jr. 2016, ACC/AHA Guideline Focused Update on Duration of Dual Antiplatelet Therapy in Patients With Coronary Artery Disease: A Report of the American College of Cardiology/American Heart Association Task Force on Clinical Practice Guidelines. J Am Coll Cardiol. 2016, 68(10), 1082-115.

Hoffman, M.K.; Goudar, S.S.; Kodkany, B.S.; Metgud, M.; Somannavar, M.; Okitawutshu, J.; Lokangaka, A.; Tshefu, A.; Bose, C.L.; Mwapule, A.; Mwenechanya, M.; Chomba, E.; Carlo, W.A.; Chicuy, J.; Figueroa, L.; Garces, A.; Krebs, N.F.; Jessani, S.; Zehra F.; Saleem, S.; Goldenberg, R.L.; Kurhe, K.; Das, P.; Patel, A.; Hibberd, P.L.; Achieng, E.; Nyongesa, P.; Esamai, F.; Liechty, E.A.; Goco, N.; Hemingway-Foday, J.; Moore, J.; Nolen, T.L.; McClure, E.M.; Koso-Thomas, M.; Miodovnik, M.; Silver, R.; Derman, R.J. ASPIRIN Study Group. Low-dose aspirin for the prevention of preterm delivery in nulliparous women with a singleton pregnancy (ASPIRIN): a randomised, double-blind, placebo-controlled trial. Lancet. 2020, 25, 395(10220), 285-293.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
Synonym aspirin, acetylsalicylic acid, 2-acetoxybenzoic acid, 2-acetyloxy benzoic acid, acetylsalicylate, o-acetylsalicylic acid, o-acetoxybenzoic acid, acylpyrin, polopiryna, easprin
PubChem CID 2244
ChEBI CHEBI:15365
IUPAC Name 2-acetyloxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 136.0°C to 140.0°C
Color White
Density 1.3500g/mL
Flash Point 250°C
Loss on Drying 0.5% max. (1 g) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula 2-(CH3CO2)C6H4CO2H
Quantity 50 g
Beilstein 10, 67
Merck Index 15, 841
Specific Gravity 1.35
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g in 5mL alcohol,17mL chloroform,,and 10-15 mL ether,slightly soluble in anhydrous ether,decomposes in boiling water,alkali
Formula Weight 180.16
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material Acetylsalicylic acid
Show More Show Less
compliance-icons
Product Identifier
  • Acetylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.