missing translation for 'onlineSavingsMsg'
Learn More
Acetylsalicylsalicylic acid, 97%, Thermo Scientific Chemicals Product Code.: 10135210

Acetylsalicylsalicylic acid, 97%, Thermo Scientific Chemicals

Product Code. 10135210
250 mg, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10135210

Brand: Thermo Scientific Acros 269352500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-75-6
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.26
MDL Number MFCD00143537
InChI Key DDSFKIFGAPZBSR-UHFFFAOYSA-N
Synonym 2-2-acetoxybenzoyl oxy benzoic acid, acetylsalicylsalicylic acid, acesalum, acetylsalicylsalicyic acid, diplosal acetate, unii-vbe72mcp5l, 2-carboxyphenyl 2-acetyloxy benzoate, diplosalsalate, ccris 1740, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl esterShow MoreShow Less
PubChem CID 10745
IUPAC Name 2-(2-acetyloxybenzoyl)oxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 145°C to 160°C
Assay Percent Range 96% min. (As is) Calculated (HPLC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 10,IV,165
Merck Index 14,1
Solubility Information Solubility in water: .. Other solubilities: soluble in alkaline solutions,soluble in alcohol,practically insoluble in benzene
Formula Weight 300.26
Percent Purity 97%
Chemical Name or Material Acetylsalicylsalicylic acid, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0454978 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number269352500,269350010
Lot NumberA0402226 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0393209 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number269352500,269350010
Lot NumberA0406903 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number269352500,269350010
Lot NumberA0408054 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
compliance-icons
Product Identifier
  • Acetylsalicylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetylsalicylsalicylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.