missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACHP, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
IκB kinase inhibitor (IC50 values are 8.5 and 250 nM for IKKβ and IKKα respectively). Selective for IKKα and IKKβ over IKK3, Syk and MAPKKK4 (IC50 values are > 20 μM). Inhibits DNA binding activity of NF-κB. Blocks NF-κB pathway in multiple myeloma cell lines; induces cell growth arrest and apoptosis.
Specifications
CAS | 406208-42-2 |
Molecular Formula | C21H24N4O2 |
Quantity | 10 mg |
Synonym | achp, 2-amino-6-2-cyclopropylmethoxy-6-hydroxyphenyl-4-4-pipe |
InChI Key | DWQVGCQPWXKRKO-JZJYNLBNSA-N |
SMILES | C1CC1COC2=CC=CC(=O)C2=C3C=C(C(=C(N3)N)C#N)C4CCNCC4 |
IUPAC Name | (6E)-2-amino-6-[2-(cyclopropylmethoxy)-6-oxocyclohexa-2,4-dien-1-ylidene]-4-piperidin-4-yl-1H-pyridine-3-carbonitrile |
Molecular Weight (g/mol) | 364.449 |
PubChem CID | 22258733 |
Formula Weight | 364.44 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'