missing translation for 'onlineSavingsMsg'
Learn More

Sherwood Scientific™ Acid Buffer

Brand:  Sherwood Scientific™ 156208

Code : 03

Additional Details : CAS Number : 64-19-7 Weight : 0.55080kg


Product Code. 12386307

  • £97.16 / 500mL
In Stock
to see stock.
Add to basket

Description

Description

Specifications

Specifications

500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COMBINED ACID BUFFER

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : acetic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers