missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ AcoustiCAL Calibrator AC-300
3M™ AcoustiCAL Calibrator AC-300 is a Type 1 calibrator for use with 3M™ Sound Level Meters and Noise Dosimeters.
Brand: 3M™ 7000042911
Code : NEW
Additional Details : Weight : 0.23810kg
Description
- Class 1 Acoustic Calibrator with two frequency levels and one sound pressure level
- Generates two settings with either 250 Hz or 1000 Hz at a sound pressure level of 114dB
Especificaciones
Dosimeter Accessory | |
250 Hz, 1000 Hz | |
114 dB |