missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation ACQUITY HSS T3 Column
The ACQUITY HSS T3 UPLC columns exhibit ultra-low MS bleed and are fully scalable and transferable to HPLC separations.
Brand: Waters Corporation 186005617
This item is not returnable.
View return policy
Description
- The universal, silica-based bonded phase used for the ACQUITY HSS T3 sorbents is compatible with 100% aqueous mobile phase and should be your first choice when developing separations for polar and non-polar compounds.
Specifications
3 mm | |
18000 psi (1240 Bar) | |
1.8 μm | |
1/Ea. | |
Yes |
75 mm | |
Spherical | |
100 Å | |
L1 | |
Column |