missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content

Product Code. 10505783 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10505783

Brand: Thermo Scientific Chemicals 423340010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1704465-79-1 | C17H20ClN3 | 301.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1704465-79-1
Molecular Formula C17H20ClN3
MDL Number MFCD00150070
Beilstein 22, 487
Synonym Basic orange 14, 3, 6-Bis(dimethylamino)acridine hydrochloride, C.I. 46005
InChI Key VSTHNGLPHBTRMB-UHFFFAOYSA-N
Infrared Spectrum Authentic
SMILES Cl.CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
IUPAC Name N3,N3,N6,N6-tetramethylacridine-3,6-diamine hydrochloride
Molecular Weight (g/mol) 301.82
PubChem CID 517204
ChEBI CHEBI:51739
Formula Weight 301.82
Loss on Drying 5% max. (105°C)
Grade Extra Pure
Packaging Glass bottle
UV_Vis 75% min.
Color Brown to Orange
Quantity 1 g
Physical Form Powder
Chemical Name or Material Acridine orange, hydrochloride hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acridine Orange, hydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.