Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PTFE Membrane
The retention efficiency of the Acrodisc PSF filter can directly increase the life of your HPLC column.
Brand:  Cytiva (formerly Pall Lab) AP-4302
Additional Details : Weight : 3.64190kg
 View more versions of this product
Product Code. 17194661
  • £2735.00 / Pack of 1000
Estimated Shipment: 05-11-2024
to see stock.
Add to basket
Description
Description
  • More retentive filters remove smaller non-dissolved particles that may rest on the head of the HPLC column and affect analytical results as they dissolve during an analysis
Specifications
Specifications
Glass Fiber, PTFE
Non-sterile
Polypropylene
100°C
0.45 μm
3.9 cm2
1000/Pk.
25 mm