missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Nylon Membrane

Versatile filter for both aqueous and solvent-based sample filtration.

Brand:  Cytiva (formerly Pall Lab) 4438

Additional Details : Weight : 1.06400kg

 View more versions of this product

Product Code. 17144311

  • £219.00 / Pack of 50
Estimated Shipment: 29-08-2024
to see stock.
Add to basket

Description

Description

  • Excellent chemical compatibility with esters, bases, and alcohols
  • Saves time because no prewetting is required
Specifications

Specifications

Nylon
Non-sterile
<125 μL
200/Cs.
55°C
0.45 μm
2.8 cm2
Polypropylene
50/Pk.
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers