missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Nylon Membrane

Versatile filter for both aqueous and solvent-based sample filtration.

Brand:  Cytiva (formerly Pall Lab) 4502

Additional Details : Weight : 3.41050kg

 View more versions of this product

Product Code. 17144331

  • £4085.00 / Pack of 1000
Estimated Shipment: 29-08-2024
to see stock.
Add to basket

Description

Description

  • Excellent chemical compatibility with esters, bases, and alcohols
  • Saves time because no prewetting is required
Specifications

Specifications

Nylon
Non-sterile
<125 μL
1000/Pk.
25 mm
0.45 μm
2.8 cm2
Polypropylene
55°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers