missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Supor™ Membrane and Modified Acrylic Housing

These syringe filters are ideal for biological and general research.

Brand:  Cytiva (formerly Pall Lab) 4659

Additional Details : Weight : 3.65750kg

 View more versions of this product

Product Code. 17184381

  • £2215.00 / Pack of 1000
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Superior flow rates and higher throughputs than competitive devices
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

PES
Non-sterile
<100 μL
1000/Pk.
32 mm
0.8/0.2 μm
5.8 cm2
Modified Acrylic
55°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers