missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Unit with Mustang E Membrane
Acrodisc™ Unit for removing endotoxins from water, buffers, neutral sugars, and certain biological solutions.
Brand: Cytiva (formerly Pall Lab) MSTG25E3
Additional Details : Weight : 0.31200kg
Description
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Highly Cross-linked Quaternized Amine Mustang E | |
Non-sterile | |
Blister Pack |
0.2 μm | |
10/Pk. | |
25 mm |