Learn More
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO)
Assess the performance of molecular test procedures for HIV-1, HCV and HBV nucleic acids with these panels, which are for Research Use Only.
£1195.00
Specifications
Packaging 7 x 1.2mL
Quantity 7 x 1.2 mL
View More Specs
Products 1
Product Code Brand Description Level Price Quantity & Availability  
Product Code Brand Description Level Price Quantity & Availability  
17633545
View Documents
Thermo Scientific™
950150
AcroMetrix HBV Panel 50, 500, 5000, 50000, 500000, 5000000, 50000000 IU/mL
£1195.00
Each
Description
Description
Assess the performance of molecular test procedures, including the extraction step, for the quantitative and qualitative determination of HIV-1, HCV and HBV nucleic acids with the Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO). These panels can be used for training and proficiency testing of laboratory personnel, for lot-to-lot comparison of reagent test kits and to evaluate and compare intralaboratory and interlaboratory assessments. The panels are for Research Use Only and are not for use in diagnostic procedures.
Specifications
Specifications
7 x 1.2mL
7 x 1.2 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ AcroMetrix™ HIV-1/HCV/HBV Panels (RUO)