Learn More
Cytiva AcroPak™ 1500 Capsules with Supor™ Membrane
AcroPak Capsules for buffers, tissue culture media, and other biological fluids.
Brand: Cytiva 12675
200.45 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
- Designed for research applications only
- Gamma-irradiated
Specifications
Hydrophilic PES | |
Sterile | |
1/2 in. Hose Barb |
0.8/0.2 μm | |
1500 cm2 | |
1/Ea. |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
3Hours
21Minutes
49Seconds
59