missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) AcroPak™ 20 Filters with Fluorodyne™ II Membrane and Filling Bell

AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations.

Brand:  Cytiva (formerly Pall Lab) 12200

Additional Details : Weight : 0.38950kg

 View more versions of this product

Product Code. 17124231

  • £92.75 / Pack of 3
Estimated Shipment: 19-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Efficient processing volumes up to 20 L
  • Fluorodyne II (PVDF) membrane is compatible with aqueous and many organic solvents
Specifications

Specifications

Hydrophilic PVDF
Non-sterile
Stepped Hose Barb
0.2 μm
20 cm2
3/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers