missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) AcroPak™ 20 Filters with Supor™ EKV Membrane

AcroPak 20 Filters are designed to add convenience to small- and medium-volume filtration (sterile and non-sterile).

Brand:  Cytiva (formerly Pall Lab) 12247

Additional Details : Weight : 0.66500kg

 View more versions of this product

Product Code. 17174231

  • £71.15 / Pack of 3
Estimated Shipment: 22-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Supor membrane has high flow rates and throughputs, and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones
  • Efficiently processes up to 2 L of chemical and biological fluids
  • Sterilization by gamma irradiation eliminates potential contamination by EtO residuals
  • Upstream vent prevents vapor lock
  • Built-in prefilter extends filter life when particulate-laden solutions such as serum-containing media are processed
Specifications

Specifications

Hydrophilic PES
Sterile
Stepped Hose Barb
0.2 μm
20 cm2
3/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers