missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 20 Filters with Supor™ Membrane and Filling Bell
Designed to add convenience to small- and medium-volume filtration (sterile and non-sterile).
£88.05 - £134.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.8/0.2 μm |
Filtration Area | 20 cm2 |
No. per Pack | 3/Pk. |
Outlet Style | Stepped Hose Barb |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
17154231
|
Cytiva (formerly Pall Lab)
12203 |
Sterile |
£134.00
Pack of 3 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17144231
|
Cytiva (formerly Pall Lab)
12202 |
Non-sterile |
£88.05
Pack of 3 |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Supor membrane has high flow rates and throughputs, and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones
- Efficiently processes up to 2 L of chemical and biological fluids
- Sterilization by gamma irradiation eliminates potential contamination by EtO residuals
- Upstream vent prevents vapor lock
- Built-in prefilter extends filter life when particulate-laden solutions such as serum-containing media are processed
- Porous membrane delivers high air flow rates at low differential pressure and extends filter life
- Ensures particulate-free air 99.97% retention of 0.3 μm DOP aerosol
- Designed for laboratory applications only
Specifications
Hydrophilic PES | |
20 cm2 | |
Stepped Hose Barb |
0.8/0.2 μm | |
3/Pk. |