Learn More
Cytiva AcroPak™ 200 Capsules with Supor™ Membrane
Click to view available options
Outlet Style:
1/4 in. MNPT Inlet, Stepped Hose Barb Outlet
Stepped Hose Barb
Unit Size:
Pack of 3
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Specifications
| Material (Membrane) | Hydrophilic PES |
| Pore Size | 0.8/0.2 μm |
| Sterility | Sterile |
| Filtration Area | 200 cm2 |
| Outlet Style | 1/4 in. MNPT Inlet, Stepped Hose Barb Outlet |
| Quantity | 3/Pk. |