Learn More
Cytiva AcroPak™ 300 Capsule with PTFE Membrane
AcroPak 300 Capsule offers broad chemical compatibility with PTFE membrane and polypropylene housing.
Brand:  Cytiva 12082
358.15 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 17114221
  • £377.00 / Pack of 3
Explore more special offers
Description
Description
  • High air flow rate venting applications
  • Offers broad chemical compatibility with PTFE membrane and polypropylene housing 100% integrity-tested to provide the high level of confidence you require
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE
Non-sterile
3/Pk.
0.2 μm
280 cm2
Stepped Hose Barb
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva AcroPak™ 300 Capsule with PTFE Membrane > 3/Pk.