missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 400 Capsules with Supor™ EKV Membrane
AcroPak capsules designed for research applications only.
£119.00
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.2 μm |
Sterility | Sterile |
Filtration Area | 375 cm2 |
Quantity | 1/Ea. |
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Outlet Style | Price | Quantity & Availability | |||||
17184231
|
Cytiva (formerly Pall Lab)
12460 |
1 to 1.5 in. Sanitary Flange |
£119.00
Each |
Estimated Shipment: 05-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17194231
|
Cytiva (formerly Pall Lab)
12461 |
1/2 in. Hose Barb Inlet with Filling Bell on Outlet |
£119.00
Each |
Estimated Shipment: 07-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
1/Ea. |
0.2 μm | |
375 cm2 |