Learn More
Cytiva (formerly Pall Lab) AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
Marke: Cytiva (formerly Pall Lab) 12991
121.60 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Spezifikation
Hydrophilic PES | |
Sterile | |
1/Ea. |
0.8/0.2 μm | |
500 cm2 | |
Stepped Hose Barb |