Learn More
Cytiva (formerly Pall Lab) AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
Brand:  Cytiva (formerly Pall Lab) 12993
Additional Details : Weight : 0.13600kg
 View more versions of this product
Product Code. 17124251
  • £234.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
Specifications
Specifications
Hydrophilic PES
Sterile
1/Ea.
0.8/0.45 μm
500 cm2
Stepped Hose Barb