Learn More
Cytiva (formerly Pall Lab) AcroPak™ 800 Capsules with Fluorodyne™ II Membrane
AcroPak 800 Capsules with Fluorodyne II (PVDF) membrane that is compatible with aqueous and many organic solvents.
Brand:  Cytiva (formerly Pall Lab) 12473
Additional Details : Weight : 0.27550kg
 View more versions of this product
Product Code. 17154241
  • £190.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Description
Description
  • Double layer sterilizing membrane enhances reliability
  • Upstream air vent prevents vapor lock
  • Manufactured without the use of adhesives to minimize extractables
  • Pre-sterilized by gamma irradiation
Specifications
Specifications
Hydrophilic PVDF
Sterile
9/16 in. Hose Barb
0.1 μm
800 cm2
1/Ea.